Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rle-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379916.1 | NP_001366782.1 | 8 - 9 | 2859 | 121 | |
NM_001379917.1 | NP_001367296.1 | 8 - 9 | 2801 | 121 | ||
NM_001379918.1 | NP_001367283.1 | 1 - 2 | 221 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |