Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: strd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027731.5 | NP_001022902.1 | 2 - 3 | 299 | 144 | |
NR_002379.1 | 2 - 3 | 273 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |