Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: obr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129258.5 | NP_001122730.1 | 3 - 4 | 498 | 112 | |
NM_001379928.1 | NP_001367300.1 | 11 - 12 | 1347 | 112 | ||
NM_001379929.1 | NP_001367172.1 | 10 - 11 | 1677 | 112 | ||
NM_001382996.1 | 11 - 12 | 1424 | 112 | |||
NM_067047.6 | NP_499448.1 | 11 - 12 | 1495 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |