Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: obr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129258.5 | NP_001122730.1 | 6 - 7 | 1106 | 57 | |
NM_001379928.1 | NP_001367300.1 | 14 - 15 | 1955 | 57 | ||
NM_001379929.1 | NP_001367172.1 | 13 - 14 | 2285 | 57 | ||
NM_001382996.1 | 14 - 15 | 2032 | 57 | |||
NM_067047.6 | NP_499448.1 | 14 - 15 | 2103 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |