Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372661.1 | NP_001359517.1 | 21 - 22 | 3256 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |