Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379936.2 | NP_001366981.1 | 4 - 5 | 368 | 75 | |
GenBank mRNA | AF031842.1 | - | 3 - 4 | 292 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |