Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027688.5 | NP_001022859.1 | 6 - 7 | 963 | 65 | |
NM_001027689.3 | NP_001022860.1 | 6 - 7 | 867 | 65 | ||
NM_001027690.3 | NP_001022861.1 | 6 - 7 | 857 | 65 | ||
NM_001027691.4 | NP_001022862.1 | 6 - 7 | 979 | 65 | ||
NM_001027692.4 | NP_001022863.1 | 6 - 7 | 864 | 65 | ||
NM_001268206.3 | NP_001255135.1 | 5 - 6 | 1170 | 65 | ||
NM_001268207.2 | NP_001255136.1 | 3 - 4 | 719 | 65 | ||
GenBank mRNA | AB081595.1 | - | 6 - 7 | 864 | 65 | |
AB081596.1 | - | 6 - 7 | 946 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |