Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067116.7 | NP_499517.1 | 2 - 3 | 242 | 72 | |
GenBank mRNA | AF326942.1 | - | 2 - 3 | 242 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |