Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd-74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067124.2 | NP_499525.1 | 4 - 5 | 725 | 83 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |