Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067125.3 | NP_499526.1 | 2 - 3 | 533 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |