Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083169.2 | NP_001076638.1 | 10 - 11 | 1504 | 119 | |
NM_001083170.3 | NP_001076639.1 | 10 - 11 | 1456 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |