Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: maa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067130.5 | NP_499531.1 | 1 - 2 | 111 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |