Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nft-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067155.8 | NP_499556.1 | 5 - 6 | 770 | 70 | |
GenBank mRNA | AF069986.1 | - | 5 - 6 | 768 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |