Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: top-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067157.6 | NP_499558.1 | 3 - 4 | 449 | 59 | |
GenBank mRNA | AF057032.1 | - | 3 - 4 | 449 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |