Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027777.4 | NP_001022948.1 | 3 - 4 | 1118 | 153 | |
NM_001027778.5 | NP_001022949.1 | 3 - 4 | 1116 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |