Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268248.4 | NP_001255177.1 | 16 - 17 | 3255 | 139 | |
NM_001268249.2 | NP_001255178.1 | 15 - 16 | 3172 | 139 | ||
NM_001312958.3 | NP_001299887.1 | 16 - 17 | 3271 | 139 | ||
NM_001312959.3 | NP_001299888.1 | 15 - 16 | 3174 | 139 | ||
NM_001312960.3 | NP_001299889.1 | 13 - 14 | 2685 | 139 | ||
NM_001312961.3 | NP_001299890.1 | 12 - 13 | 2595 | 139 | ||
NM_001312962.3 | NP_001299891.1 | 13 - 14 | 2676 | 139 | ||
NM_001312963.3 | NP_001299892.1 | 12 - 13 | 2586 | 139 | ||
NM_001312964.3 | NP_001299893.1 | 12 - 13 | 2115 | 139 | ||
NM_001312965.3 | NP_001299894.1 | 11 - 12 | 2025 | 139 | ||
NM_001312966.3 | NP_001299895.1 | 12 - 13 | 2106 | 139 | ||
NM_001312967.3 | NP_001299896.1 | 11 - 12 | 2016 | 139 | ||
NM_001312968.3 | NP_001299897.1 | 3 - 4 | 687 | 139 | ||
NM_001312969.3 | NP_001299898.1 | 2 - 3 | 597 | 139 | ||
NM_001312970.3 | NP_001299899.1 | 1 - 2 | 78 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |