Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268248.4 | NP_001255177.1 | 21 - 22 | 4278 | 67 | |
NM_001268249.2 | NP_001255178.1 | 20 - 21 | 4195 | 67 | ||
NM_001312958.3 | NP_001299887.1 | 21 - 22 | 4294 | 67 | ||
NM_001312959.3 | NP_001299888.1 | 20 | 4197 | 67 | ||
NM_001312960.3 | NP_001299889.1 | 18 - 19 | 3708 | 67 | ||
NM_001312961.3 | NP_001299890.1 | 17 - 18 | 3618 | 67 | ||
NM_001312962.3 | NP_001299891.1 | 18 - 19 | 3699 | 67 | ||
NM_001312963.3 | NP_001299892.1 | 17 - 18 | 3609 | 67 | ||
NM_001312964.3 | NP_001299893.1 | 17 - 18 | 3138 | 67 | ||
NM_001312965.3 | NP_001299894.1 | 16 - 17 | 3048 | 67 | ||
NM_001312966.3 | NP_001299895.1 | 17 - 18 | 3129 | 67 | ||
NM_001312967.3 | NP_001299896.1 | 16 - 17 | 3039 | 67 | ||
NM_001312968.3 | NP_001299897.1 | 8 - 9 | 1710 | 67 | ||
NM_001312969.3 | NP_001299898.1 | 7 - 8 | 1620 | 67 | ||
NM_001312970.3 | NP_001299899.1 | 6 - 7 | 1101 | 67 | ||
NM_001379963.1 | NP_001368536.1 | 3 - 4 | 456 | 67 | ||
NM_001379964.1 | NP_001368537.1 | 2 - 3 | 273 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |