Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268248.4 | NP_001255177.1 | 2 - 3 | 225 | 94 | |
NM_001268249.2 | NP_001255178.1 | 2 - 3 | 232 | 94 | ||
NM_001312958.3 | NP_001299887.1 | 2 - 3 | 232 | 94 | ||
NM_001312959.3 | NP_001299888.1 | 2 - 3 | 225 | 94 | ||
NM_001372667.1 | NP_001360800.1 | 2 - 3 | 232 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |