Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tpst-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067245.5 | NP_499646.3 | 2 - 3 | 306 | 121 | |
GenBank mRNA | AF049709.1 | - | 2 - 3 | 289 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |