Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ssl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268250.2 | NP_001255179.1 | 2 - 3 | 413 | 87 | |
NM_001268251.3 | NP_001255180.1 | 2 - 3 | 391 | 87 | ||
GenBank mRNA | AY551965.1 | - | 2 - 3 | 435 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |