Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ssl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268250.2 | NP_001255179.1 | 5 - 6 | 2059 | 116 | |
NM_001268251.3 | NP_001255180.1 | 5 - 6 | 2037 | 116 | ||
NM_001379967.1 | NP_001367266.1 | 3 - 4 | 1599 | 116 | ||
GenBank mRNA | AY551965.1 | - | 5 - 6 | 2081 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |