Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027660.3 | NP_001022831.1 | 5 - 6 | 940 | 67 | |
NM_001027661.5 | NP_001022832.1 | 3 - 4 | 423 | 67 | ||
NM_067288.5 | NP_499689.1 | 5 - 6 | 656 | 67 | ||
GenBank mRNA | AF109378.1 | - | 5 - 6 | 618 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |