Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379972.2 | NP_001367103.1 | 6 - 7 | 1057 | 158 | |
NM_001383018.1 | 6 - 7 | 1061 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |