Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zig-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067313.7 | NP_499714.1 | 4 - 5 | 481 | 71 | |
GenBank mRNA | AF456253.1 | - | 4 - 5 | 442 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |