Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zig-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067313.7 | NP_499714.1 | 5 - 6 | 617 | 82 | |
GenBank mRNA | AF456253.1 | - | 5 - 6 | 578 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |