Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bre-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268259.3 | NP_001255188.1 | 5 - 6 | 631 | 166 | |
NM_001268260.1 | NP_001255189.1 | 5 - 6 | 619 | 166 | ||
NM_001379974.2 | NP_001367175.1 | 3 - 4 | 496 | 166 | ||
NM_067314.4 | NP_499715.2 | 5 - 6 | 631 | 166 | ||
GenBank mRNA | AY533304.1 | - | 3 - 4 | 496 | 166 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |