Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pssy-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067385.6 | NP_499786.3 | 8 - 9 | 1222 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |