Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268276.2 | NP_001255205.1 | 3 - 4 | 444 | 105 | |
NM_001268277.1 | NP_001255206.1 | 3 - 4 | 460 | 105 | ||
NM_001268278.2 | NP_001255207.1 | 2 - 3 | 331 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |