Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pod-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276804.3 | NP_001263733.1 | 4 - 5 | 2651 | 81 | |
NM_001276805.4 | NP_001263734.1 | 5 - 6 | 2784 | 81 | ||
NM_001276806.2 | NP_001263735.1 | 5 - 6 | 2772 | 81 | ||
NM_067396.6 | NP_499797.1 | 4 - 5 | 2640 | 81 | ||
GenBank mRNA | AF129126.2 | - | 4 - 5 | 2662 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |