Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osm-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268280.2 | NP_001255209.1 | 5 - 6 | 845 | 88 | |
NM_001268281.3 | NP_001255210.1 | 5 - 6 | 829 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |