Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083186.4 | NP_001076655.1 | 3 - 4 | 361 | 103 | |
GenBank mRNA | DQ314286.1 | - | 3 - 4 | 361 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |