Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083186.4 | NP_001076655.1 | 4 - 5 | 558 | 99 | |
GenBank mRNA | DQ314286.1 | - | 4 - 5 | 558 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |