Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083186.4 | NP_001076655.1 | 5 - 6 | 955 | 72 | |
GenBank mRNA | DQ314286.1 | - | 5 - 6 | 955 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |