Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scl-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067458.5 | NP_499859.3 | 5 - 6 | 707 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |