Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: col-99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129303.4 | NP_001122775.2 | 11 - 12 | 1321 | 74 | |
NM_001307632.3 | NP_001294561.1 | 9 - 10 | 1258 | 74 | ||
NM_001307634.4 | NP_001294563.1 | 10 - 11 | 1294 | 74 | ||
NM_001322766.3 | NP_001309610.1 | 9 - 10 | 1258 | 74 | ||
NM_001322767.3 | NP_001309606.1 | 9 - 10 | 1258 | 74 | ||
NM_001322770.3 | NP_001309609.1 | 10 - 11 | 1294 | 74 | ||
NM_001322771.3 | NP_001309605.1 | 10 - 11 | 1294 | 74 | ||
NM_001322774.3 | NP_001309603.1 | 9 - 10 | 1258 | 74 | ||
NM_067468.7 | NP_499869.3 | 10 - 11 | 1294 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |