Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028278.5 | NP_001023449.1 | 5 - 6 | 2003 | 85 | |
NM_001028279.6 | NP_001023450.1 | 3 - 4 | 434 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |