Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dsc-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067502.6 | NP_499903.3 | 4 - 5 | 889 | 68 | |
GenBank mRNA | AY428645.1 | - | 4 - 5 | 923 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |