Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdh-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307480.3 | NP_001294409.1 | 22 - 23 | 4461 | 86 | |
NM_001307481.3 | NP_001294410.1 | 21 - 22 | 4266 | 86 | ||
NM_001307482.3 | NP_001294411.1 | 22 - 23 | 4359 | 86 | ||
NM_001307483.3 | NP_001294412.1 | 23 - 24 | 4554 | 86 | ||
NM_001307484.3 | NP_001294413.1 | 23 - 24 | 4416 | 86 | ||
NM_001307485.3 | NP_001294414.1 | 22 - 23 | 4221 | 86 | ||
NM_001307486.3 | NP_001294415.1 | 23 - 24 | 4314 | 86 | ||
NM_001307487.3 | NP_001294416.1 | 24 - 25 | 4509 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |