Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307363.3 | NP_001294292.1 | 18 - 19 | 2055 | 125 | |
NM_001307364.3 | NP_001294293.1 | 17 - 18 | 2061 | 125 | ||
NM_001307365.3 | NP_001294294.1 | 16 - 17 | 1938 | 125 | ||
NM_068737.2 | NP_501138.1 | 18 - 19 | 2058 | 125 | ||
GenBank mRNA | U76402.1 | - | 18 - 19 | 2058 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |