Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kcc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306917.3 | NP_001293846.1 | 9 - 10 | 1227 | 100 | |
NM_001306918.3 | NP_001293847.1 | 8 - 9 | 1053 | 100 | ||
NM_001306919.3 | NP_001293848.1 | 7 - 8 | 957 | 100 | ||
NM_001307143.2 | NP_001294072.1 | 6 - 7 | 1092 | 100 | ||
NM_001380315.1 | NP_001368174.1 | 6 - 7 | 864 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |