Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068741.2 | NP_501142.1 | 5 - 6 | 936 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |