Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plk-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068795.7 | NP_501196.1 | 4 - 5 | 1111 | 151 | |
GenBank mRNA | AF059024.1 | - | 4 - 5 | 1111 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |