Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pam-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028038.7 | NP_001023209.1 | 5 - 6 | 1437 | 104 | |
NM_001028039.3 | NP_001023210.1 | 4 - 5 | 937 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |