Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cla-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307214.3 | NP_001294143.1 | 13 - 14 | 2509 | 123 | |
NM_001307215.3 | NP_001294144.1 | 13 - 14 | 2509 | 123 | ||
NM_001307219.4 | NP_001294148.2 | 26 - 27 | 21349 | 123 | ||
NM_001307220.4 | NP_001294149.2 | 26 - 27 | 21349 | 123 | ||
NM_001383125.1 | 6 - 7 | 1839 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |