Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cla-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307214.3 | NP_001294143.1 | 5 - 6 | 572 | 87 | |
NM_001307215.3 | NP_001294144.1 | 5 - 6 | 572 | 87 | ||
NM_001307219.4 | NP_001294148.2 | 18 - 19 | 19412 | 87 | ||
NM_001307220.4 | NP_001294149.2 | 18 - 19 | 19412 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |