Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nep-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028032.6 | NP_001023203.2 | 10 - 11 | 1394 | 100 | |
NM_001028034.5 | NP_001023205.2 | 8 - 9 | 1058 | 100 | ||
NM_001383127.1 | 9 - 10 | 1190 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |