Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: itr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027999.4 | NP_001023170.1 | 29 - 30 | 7425 | 83 | |
NM_001028000.5 | NP_001023171.1 | 27 - 28 | 7257 | 83 | ||
NM_001028001.5 | NP_001023172.1 | 29 - 30 | 7388 | 83 | ||
NM_001028002.4 | NP_001023173.1 | 28 - 29 | 7353 | 83 | ||
NM_001268376.4 | NP_001255305.1 | 29 - 30 | 7350 | 83 | ||
GenBank mRNA | AF168688.1 | - | 28 - 29 | 7353 | 83 | |
AJ243179.2 | - | 28 - 29 | 7375 | 83 | ||
AJ243180.2 | - | 29 - 30 | 7448 | 83 | ||
AJ243181.2 | - | 28 - 29 | 7368 | 83 | ||
AJ243182.2 | - | 29 - 30 | 7408 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |