Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mvb-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068901.7 | NP_501302.1 | 4 - 5 | 464 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |