Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xpo-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307224.3 | NP_001294153.1 | 9 - 10 | 1385 | 109 | |
NM_001380346.1 | NP_001368437.1 | 2 - 3 | 404 | 109 | ||
NM_068919.8 | NP_501320.2 | 9 - 10 | 1379 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |