Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bbs-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068924.3 | NP_501325.2 | 8 - 9 | 1990 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |